Learn More
Palintest™ Zinc Starter Tdisc Test
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
Description
The Palintest™ comparator with interchangeable colour discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.
Specifications
Specifications
| Concentration | 0 to 4 mg/L |
| Quantity | 50 Pack |