missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium(IV) 2-ethylhexanoate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Zirconium(IV) 2-ethylhexanoate is utilized as a metal organic precursor of zirconium for solution deposition and chemical vapor deposition of thin films containing zirconium especially zirconium oxide thin films. It is also used as yttria stabilized zirconia films for solid oxide fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Amber |
Flash Point | >93°C (199°F) |
Quantity | 25 g |
Sensitivity | Moisture sensitive |
Formula Weight | 664.04 |
Percent Purity | 97% |
Physical Form | Viscous Liquid |
Chemical Name or Material | Zirconium(IV) 2-ethylhexanoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'