Learn More
RETSCH Zirconium Oxide Grinding Jars
Shop All Retsch Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
For CryoMill
For EMAX
For MM 200 Mixer Mill
For MM 301 Mixer mill
Capacity (Metric):
10 mL
25 mL
50 mL
125 mL
missing translation for 'unitSize'
Each
Description
- Accessory
Specifications
Specifications
| Product Type | Grinding Jar |
| Capacity (Metric) | 10 mL |
| For Use With (Equipment) | For MM 200 Mixer Mill |
| Material | Zirconium Oxide |