missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ ZLD 1039

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ZLD 1039 is a potent and selective EZH2 inhibitor (IC50 = 5.6 nM). Displays selectivity for EZH2 over a panel of HMTs (inhibition less than 30% at 16 μM). Blocks H3K27 methylation in breast cancer cell lines. Induces tumor regression and metastasis in breast cancer xenograft-bearing mice. Reactivates silenced tumor suppressors in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 3-[Ethyl(tetrahydro-2H-pyran-4-yl)amino]-N-[(2,3,5,6,7,8-hexahydro-1-methyl-3-oxo-4-isoquinolinyl)methyl]-2-methyl-5-[6-(4-methyl-1-piperazinyl)-3-pyridinyl]benzamide |
| CAS | 1826865-46-6 |
| Quantity | 10 mg |
| Molecular Formula | C36H48N6O3 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction