missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Gene ID (Entrez) | 10269 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB 100-2388 |
| Gene Alias | zona pellucida binding protein 2, zona pellucida-binding protein 2, ZPBP-like protein, ZPBPLMGC41930 |
| Gene Symbol | ZMPSTE24 |
| Quantity | 0.1 mg |
| Research Category | Angiogenesis, Cancer, Cell Biology, Cellular Markers, Signal Transduction, Ubiquitin Proteasome Pathway |
| Storage Buffer | Peptide dissolved in dH2O. Contains no BSA. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?