missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ 92-134 Nitrile Glove

MICROFLEX™ 92-134 Nitrile Gloves offer tactility and comfortable protection across numerous applications. Designed with textured fingertips for secure handling of tools and materials.

Brand:  Ansell 92134090

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 17823692

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Description

Description

  • Made of a soft nitrile formulation for comfortable fit and feel
  • Lightweight construction for tactile sensitivity and fine motor control
  • Designed for a broad range of applications across all industries
  • Textured on the fingertips for secure handling of tools and materials 
Specifications

Specifications

Healthcare
Blue
Partially Textured
0.11 mm
Nitrile
0.07 mm
Large
1.5
Single-use
4.3 mil
Yes
2.8 mil
Powder Free
100 gloves/dispenser, 10 dispensers/Cs., 1000 gloves/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions