Learn More
Ansell MICROFLEX™ 92-134 Nitrile Glove
MICROFLEX™ 92-134 Nitrile Gloves offer tactility and comfortable protection across numerous applications. Designed with textured fingertips for secure handling of tools and materials.
8.50€ - 13.86€
Specifications
Applications | Healthcare |
---|---|
AQL | 1.5 |
Color | Blue |
Disposable | Single-use |
Finger Thickness (Metric) | 0.11 mm |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17601036
|
Ansell
92134060 |
X-Small |
8.50€
Pack of 100 |
In Stock
Log in to see stock. |
|||||
17611036
|
Ansell
92134070 |
Small |
8.50€
Pack of 100 |
In Stock
Log in to see stock. |
|||||
17621036
|
Ansell
92134080 |
Medium |
8.50€
Pack of 100 |
In Stock
Log in to see stock. |
|||||
17631036
|
Ansell
92134090 |
Large |
8.50€
Pack of 100 |
In Stock
Log in to see stock. |
|||||
17818331
|
Ansell
92134110 |
10.5/11, 2X-Large |
8.50€
Pack of 90 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
17641036
|
Ansell
92134100 |
X-Large |
8.50€
Pack of 100 |
In Stock
Log in to see stock. |
|||||
17853792
|
Ansell
92134080 |
Medium |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
17833612
|
Ansell
92134070 |
Small |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
17823692
|
Ansell
92134090 |
Large |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Made of a soft nitrile formulation for comfortable fit and feel
- Lightweight construction for tactile sensitivity and fine motor control
- Designed for a broad range of applications across all industries
- Textured on the fingertips for secure handling of tools and materials
Specifications
Healthcare | |
Blue | |
0.11 mm | |
Nitrile | |
0.07 mm |
1.5 | |
Single-use | |
Yes | |
2.8 mil | |
Powder Free |