accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSN

CAS RN 1003-32-3

IUPAC Name: 1,3-thiazole-5-carbaldehyde
Synonyms: 1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 113.13
Molecular Formula: C4H3NOS
InChi Key: ZXRLWHGLEJGMNO-UHFFFAOYSA-N
SMILES: O=CC1=CN=CS1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results