accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CNH3COH

CAS RN 100442-33-9

IUPAC Name: 1-[(3,3-diphenylpropyl)(methyl)amino]-2-methylpropan-2-ol
Synonyms: 1-[(3,3-diphenylpropyl)(methyl)amino]-2-methylpropan-2-ol
Molecular Weight (g/mol): 297.44
Molecular Formula: C20H27NO
InChi Key: MQWDISMNBYOLAB-UHFFFAOYSA-N
SMILES: CN(CCC(C1=CC=CC=C1)C1=CC=CC=C1)CC(C)(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results