accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HC

CAS RN 10147-11-2

IUPAC Name: (prop-2-yn-1-yl)benzene
Synonyms: prop-2-yn-1-ylbenzene
Molecular Weight (g/mol): 116.16
Molecular Formula: C9H8
InChi Key: NGKSKVYWPINGLI-UHFFFAOYSA-N
SMILES: C#CCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)