missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropyne, TRC

Product Code. 30426411
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
10g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426411 10 g 10g
30418711 2.5 g 2.5g
30422786 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426411 Supplier Toronto Research Chemicals Supplier No. TRCP33629010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10147-11-2
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
Synonym 2-Propyn-1-yl-benzene, (2-Propynyl)-benzene, 2-Propynyl-benzene, 3-Phenyl- propyne, 1-Phenyl-2-propyne, 2-Propynylbenzene, 3-Phenyl-1-propyne, Benzylacetylene, Propargylbenzene
IUPAC Name prop-2-ynylbenzene
SMILES C#CCc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H8/c1-2-6-9-7-4-3-5-8-9/h1,3-5,7-8H,6H2
Formula Weight 116.0626
Chemical Name or Material 3-Phenylpropyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.