CAS RN 1015855-83-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ipronidazole-d3 (1-methyl-d3), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Ipronidazole-d3, TRC
CAS: 1015855-83-0 Molecular Formula: C7 2H3 H8 N3 O2 Molecular Weight (g/mol): 172.2 Synonym: 1H-Imidazole, 1-(methyl-d3)-2-(1-methylethyl)-5-nitro- (ACI),1-(Methyl-d3)-2-(1-methylethyl)-5-nitro-1H-imidazole (ACI),Ipronidazole D3,Ipronidazole-D3 (N-methyl-D3),Ipronidazole-d3 IUPAC Name: 5-nitro-2-propan-2-yl-1-(trideuteriomethyl)imidazole SMILES: [2H]C([2H])([2H])n1c(ncc1[N+](=O)[O-])C(C)C