accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBr

CAS RN 10160-24-4

IUPAC Name: 7-bromoheptan-1-ol
Synonyms: 7-bromoheptan-1-ol
Molecular Weight (g/mol): 195.1
Molecular Formula: C7H15BrO
InChi Key: MMXRRNUXCHUHOE-UHFFFAOYSA-N
SMILES: OCCCCCCCBr
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)