accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1017777-45-5

H3COFClF

CAS RN 1017777-45-5

IUPAC Name: 1-(4-chloro-2,6-difluorophenyl)ethan-1-one
Synonyms: 1-(4-chloro-2,6-difluorophenyl)ethanone
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: VEZAUYYYYPJSKR-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results