CAS RN 101988-70-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Methyl-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine-2,4-diamine, TRC
CAS: 101988-70-9 Molecular Formula: C6H8F3N5O Molecular Weight (g/mol): 223.16 Synonym: Triflusulfuron-methyl Metabolite IN-E7710,1,3,5-Triazine-2,4-diamine, N-methyl-6-(2,2,2-trifluoroethoxy)- (9CI),N2-Methyl-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine-2,4-diamine (ACI),2-N-Methyl-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine-2,4-diamine,1,3,5-Triazine-2,4-diamine, N2-methyl-6-(2,2,2-trifluoroethoxy)- (ACI) IUPAC Name: 2-N-methyl-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine-2,4-diamine SMILES: CNc1nc(N)nc(OCC(F)(F)F)n1