missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine-2,4-diamine, TRC

Product Code. 30133788
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133788 500 mg 500mg
30012640 50 mg 50mg
30011665 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133788 Supplier Toronto Research Chemicals Supplier No. TRCM330478500MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101988-70-9
Molecular Formula C6H8F3N5O
Molecular Weight (g/mol) 223.16
Synonym Triflusulfuron-methyl Metabolite IN-E7710, 1,3,5-Triazine-2,4-diamine, N-methyl-6-(2,2,2-trifluoroethoxy)- (9CI), N2-Methyl-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine-2,4-diamine (ACI), 2-N-Methyl-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine-2,4-diamine, 1,3,5-Triazine-2,4-diamine, N2-methyl-6-(2,2,2-trifluoroethoxy)- (ACI)
IUPAC Name 2-N-methyl-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine-2,4-diamine
SMILES CNc1nc(N)nc(OCC(F)(F)F)n1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H8F3N5O/c1-11-4-12-3(10)13-5(14-4)15-2-6(7,8)9/h2H2,1H3,(H3,10,11,12,13,14)
Formula Weight 223.0681
Chemical Name or Material N-Methyl-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine-2,4-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.