man

Test

CAS RN 10265-92-6

CH3OPNH2OSH3C

CAS RN 10265-92-6

IUPAC Name: [methoxy(methylsulfanyl)phosphoryl]amine
Synonyms: tamaron sniper methamidophos chevron 9006 chevron ortho 9006 patrole ortho monitor MTD methyl phosphoramidothioate epa pesticide chemical code 101201
Molecular Weight (g/mol): 141.12
Molecular Formula: C2H8NO2PS
InChi Key: NNKVPIKMPCQWCG-UHFFFAOYNA-N
SMILES: COP(N)(=O)SC
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (3)
  • (2)
  • (1)
Color 
  • (1)
For Use With (Equipment) 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)