missing translation for 'onlineSavingsMsg'
Learn More

Metamidophos (>90%), TRC

Product Code. 30041040
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041040 25 mg 25mg
30040548 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30041040 Supplier Toronto Research Chemicals Supplier No. TRCM25884025MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10265-92-6
Molecular Formula C2 H8 N O2 P S
Molecular Weight (g/mol) 141.13
Synonym Methamidophos
IUPAC Name [amino(methylsulfanyl)phosphoryl]oxymethane
SMILES COP(=O)(N)SC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C2H8NO2PS/c1-5-6(3,4)7-2/h1-2H3,(H2,3,4)
Formula Weight 141.0013
Chemical Name or Material Methamidophos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.