accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 103260-65-7

CH3OHNOHO

CAS RN 103260-65-7

IUPAC Name: 4-methoxy-1H-indole-2-carboxylic acid
Synonyms: 4-methoxy-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: ZZAVIQXQBBOHBB-UHFFFAOYSA-N
SMILES: COC1=C2C=C(NC2=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results