CAS RN 103404-90-6
CAS RN 103404-90-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-2-Hydroxyglutaric acid disodium salt, Tocris Bioscience™
KDM4A inhibitor; also NF-κB activator