missing translation for 'onlineSavingsMsg'
Learn More

D-alpha-Hydroxyglutaric Acid Disodium, 95%, BLD Pharm™

Product Code. 31029778
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
5 g
Unit Size:
1g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31029778 100 mg -
31021773 1 g 1g
31018114 5 g -
31018826 250 mg -
31021697 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31029778 Supplier BLD Pharm Supplier No. BD123298100mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 103404-90-6
Molecular Formula C5H6Na2O5
MDL Number MFCD00069573
Quantity 100 mg
SMILES O=C([O-])[C@H](O)CCC([O-])=O.[Na+].[Na+]
Molecular Weight (g/mol) 192.08
Percent Purity 95%
Chemical Name or Material D-alpha-Hydroxyglutaric Acid Disodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.