accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH

CAS RN 10354-00-4

IUPAC Name: tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ol
Synonyms: tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ol
Molecular Weight (g/mol): 208.26
Molecular Formula: C15H12O
InChi Key: SRIISEYIFDTFRZ-UHFFFAOYSA-N
SMILES: OC1C2=CC=CC=C2C=CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)