accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO(R)OH

CAS RN 103548-16-9

IUPAC Name: (1R)-1-phenylpropane-1,3-diol
Synonyms: (1R)-1-phenylpropane-1,3-diol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: RRVFYOSEKOTFOG-SECBINFHSA-N
SMILES: OCC[C@@H](O)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results