CAS RN 104561-41-3
CAS RN 104561-41-3
IUPAC Name:
4-[2-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethynyl]benzoic acid
Synonyms:
4-[2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethynyl]benzoic acid
Molecular Weight (g/mol):
332.44
Molecular Formula:
C23H24O2
InChi Key:
OQVLOWLEEHYBJH-UHFFFAOYSA-N
SMILES:
CC1(C)CCC(C)(C)C2=C1C=CC(=C2)C#CC1=CC=C(C=C1)C(O)=O
1
–
2
of
2
results
1
Tocris Bioscience™ EC 23

CAS: 104561-41-3 Molecular Formula: C23H24O2 Molecular Weight (g/mol): 332.443
2