accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOOBr

CAS RN 104777-39-1

IUPAC Name: 2-bromo-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)ethan-1-one
Synonyms: 2-bromo-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)ethanone
Molecular Weight (g/mol): 280.12
Molecular Formula: C12H10BrNO2
InChi Key: QKOOGOQWNSWJFQ-UHFFFAOYSA-N
SMILES: CC1=C(C(=O)CBr)C(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results