accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 105-54-4

IUPAC Name: ethyl butanoate
Synonyms: ethyl butyrate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: OBNCKNCVKJNDBV-UHFFFAOYSA-N
SMILES: CCCC(=O)OCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results