missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Butyrate, TRC

Product Code. 30377961
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377961 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377961 Supplier Toronto Research Chemicals Supplier No. TRCE9006905G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butyric acid ethyl ester
CAS 105-54-4
Molecular Formula C6 H12 O2
Quantity 5 g
Synonym Butyric acid, ester with EtOH (4CI), Butyric acid, ethyl ester (6CI,8CI), Ethyl butanoate, Ethyl butyrate, Ethyl n-butanoate, Ethyl n-butyrate, NSC 8857
InChI Formula InChI=1S/C6H12O2/c1-3-5-6(7)8-4-2/h3-5H2,1-2H3
SMILES CCCC(=O)OCC
IUPAC Name ethyl butanoate
Molecular Weight (g/mol) 116.16
Formula Weight 116.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.