CAS RN 10519-33-2
CAS RN 10519-33-2
IUPAC Name:
(3E)-dec-3-en-2-one
Synonyms:
(3E)-dec-3-en-2-one
Molecular Weight (g/mol):
154.25
Molecular Formula:
C10H18O
InChi Key:
JRPDANVNRUIUAB-CMDGGOBGSA-N
SMILES:
CCCCCC\C=C\C(C)=O
1
–
2
of
2
results
1
3-Decen-2-one, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
3-Decen-2-one, 97%, Thermo Scientific™
CAS: 10519-33-2 Molecular Formula: C10H18O Molecular Weight (g/mol): 154.253 MDL Number: MFCD00015700 InChI Key: JRPDANVNRUIUAB-CMDGGOBGSA-N Synonym: 3-decen-2-one,heptylidene acetone,oenanthylidene acetone,e-3-decen-2-one,unii-z22804bqxd,3e-dec-3-en-2-one,e-dec-3-en-2-one,dec-3-en-2-one,fema no. 3532,enanthylidene acetone PubChem CID: 5363233 IUPAC Name: (E)-dec-3-en-2-one SMILES: CCCCCCC=CC(=O)C