accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 105628-07-7

HClOSONHNN

CAS RN 105628-07-7

IUPAC Name: hydrogen 5-(1,4-diazepane-1-sulfonyl)isoquinoline chloride
Synonyms: hydrogen fasudil chloride
Molecular Weight (g/mol): 327.83
Molecular Formula: C14H18ClN3O2S
InChi Key: LFVPBERIVUNMGV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].O=S(=O)(N1CCCNCC1)C1=C2C=CN=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)