missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fasudil Monohydrochloride Salt, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 1-(5-Isoquinolinesulfonyl)-1H-hexahydro-1,4-diazepine, Hydrochloride |
| CAS | 105628-07-7 |
| Molecular Formula | C14 H17 N3 O2 S . Cl H |
| Quantity | 250 mg |
| Synonym | Isoquinoline, 5-[(hexahydro-1H-1,4-diazepin-1-yl)sulfonyl]-, hydrochloride (1:1), 1H-1,4-Diazepine, hexahydro-1-(5-isoquinolinylsulfonyl)-, monohydrochloride (9CI), 1-(5-Isoquinolinesulfonyl)homopiperazine hydrochloride, Eril, Fasudil hydrochloride |
| InChI Formula | InChI=1S/C14H17N3O2S.ClH/c18-20(19,17-9-2-6-15-8-10-17)14-4-1-3-12-11-16-7-5-13(12)14;/h1,3-5,7,11,15H,2,6,8-10H2;1H |
| SMILES | Cl.O=S(=O)(N1CCCNCC1)c2cccc3cnccc23 |
| IUPAC Name | 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride |
| Molecular Weight (g/mol) | 327.83 |
| Formula Weight | 327.0808 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction