CAS RN 10575-25-4
IUPAC Name:
4-(diethylamino)but-2-yn-1-ol
Synonyms:
4-(diethylamino)but-2-yn-1-ol
Molecular Weight (g/mol):
141.21
Molecular Formula:
C8H15NO
InChi Key:
ACGZBRWTWOZSFU-UHFFFAOYSA-N
SMILES:
CCN(CC)CC#CCO
1
–
3
of
3
results
1
4,4-Diethylamino-2-butyn-1-ol, 98%
CAS: 10575-25-4 Molecular Formula: C8H15NO Molecular Weight (g/mol): 141.21 MDL Number: MFCD00671356 InChI Key: ACGZBRWTWOZSFU-UHFFFAOYSA-N PubChem CID: 82735 IUPAC Name: 4-(diethylamino)but-2-yn-1-ol SMILES: CCN(CC)CC#CCO
2
4-(Diethylamino)-2-butyn-1-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Diethylaminobut-2-yn-1-ol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.