missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylaminobut-2-yn-1-ol, Mikromol™

Product Code. 30050389
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050389 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050389 Supplier Mikromol Supplier No. MM0047.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Diethylaminobut-2-yn-1-ol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Diethylaminobut-2-yn-1-ol
CAS 10575-25-4
Analyte or Component Names 4-Diethylaminobut-2-yn-1-ol
Shipping Condition Room Temperature
Molecular Formula C8 H15 N O
Certifications/Compliance ISO 9001
Synonym 4-Diethylaminobut-2-yn-1-ol
InChI Formula InChI=1S/C8H15NO/c1-3-9(4-2)7-5-6-8-10/h10H,3-4,7-8H2,1-2H3
SMILES CCN(CC)CC#CCO
IUPAC Name 4-(diethylamino)but-2-yn-1-ol
Molecular Weight (g/mol) 141.21
Quantity 100 mg
Formula Weight 141.1154 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.