accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10603-45-9

CH3NBr

CAS RN 10603-45-9

IUPAC Name: 3-bromo-1-methylpyrrolidine
Synonyms: 3-bromo-1-methylpyrrolidine
Molecular Weight (g/mol): 164.05
Molecular Formula: C5H10BrN
InChi Key: ZHUYTHOKJZSZQJ-UHFFFAOYNA-N
SMILES: CN1CCC(Br)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results