accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 107-45-9

H3CCH3H3CCH3CH3H2N

CAS RN 107-45-9

IUPAC Name: 2,4,4-trimethylpentan-2-amine
Synonyms: 2,4,4-trimethylpentan-2-amine
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: QIJIUJYANDSEKG-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results