missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethylpentan-2-amine (2-Amino-2,4,4-trimethylpentane; 1,1,3,3-Tetramethylbutylamine), Mikromol™

Product Code. 30050054
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050054 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050054 Supplier Mikromol Supplier No. MM0273.150100
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,4,4-Trimethylpentan-2-amine (2-Amino-2,4,4-trimethylpentane; 1,1,3,3-Tetramethylbutylamine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Octylamine
CAS 107-45-9
Analyte or Component Names tert-Octylamine
Shipping Condition Room Temperature
Molecular Formula C8 H19 N
Certifications/Compliance ISO 9001
Synonym 2-Pentanamine, 2,4,4-trimethyl-, Butylamine, 1,1,3,3-tetramethyl- (6CI,7CI,8CI), 2,4,4-Trimethyl-2-pentanamine, 1,1,3,3-Tetramethylbutanamine, 1,1,3,3-Tetramethylbutylamine, 2,4,4-Trimethyl-2-pentylamine, 2-Amino-2,4,4-trimethylpentane, NSC 33852, Primene TOA, tert-Octanamine, tert-Octylamine, 2,4,4-Trimethylpentan-2-amine
InChI Formula InChI=1S/C8H19N/c1-7(2,3)6-8(4,5)9/h6,9H2,1-5H3
SMILES CC(C)(C)CC(C)(C)N
IUPAC Name 2,4,4-trimethylpentan-2-amine
Molecular Weight (g/mol) 129.24
Quantity 100 mg
Formula Weight 129.1517 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.