accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1073355-05-1

BOOH3CCH3H3CH3C

CAS RN 1073355-05-1

IUPAC Name: 2-(3-benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3-benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 294.2
Molecular Formula: C19H23BO2
InChi Key: VNZSTZMLJUPNQM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(CC2=CC=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results