accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHN

CAS RN 1074-86-8

IUPAC Name: 1H-indole-4-carbaldehyde
Synonyms: 1H-indole-4-carbaldehyde
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: JFDDFGLNZWNJTK-UHFFFAOYSA-N
SMILES: O=CC1=C2C=CNC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results