accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOCH3

CAS RN 108-59-8

IUPAC Name: 1,3-dimethyl propanedioate
Synonyms: dimethyl malonate
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: BEPAFCGSDWSTEL-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)