accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3N

CAS RN 108-99-6

IUPAC Name: 3-methylpyridine
Synonyms: 3-methylpyridine 3-mepy
Molecular Weight (g/mol): 93.13
Molecular Formula: C6H7N
InChi Key: ITQTTZVARXURQS-UHFFFAOYSA-N
SMILES: CC1=CC=CN=C1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (4)