missing translation for 'onlineSavingsMsg'
Learn More

3-Picoline, TRC

Product Code. 30421497
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421497 50 g
30414387 1 g
30415289 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421497 Supplier Toronto Research Chemicals Supplier No. TRCP43720050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-99-6
Molecular Formula C6 H7 N
Molecular Weight (g/mol) 93.13
Synonym 3-Picoline (8CI), 3-Methylpyridine, NSC 18251, m-Methylpyridine, m-Picoline, β-Methylpyridine, β-Picoline
IUPAC Name 3-methylpyridine
SMILES Cc1cccnc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H7N/c1-6-3-2-4-7-5-6/h2-5H,1H3
Formula Weight 93.0578
Chemical Name or Material 3-Picoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.