accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1080-44-0

IUPAC Name: 2-(4-methylbenzenesulfonamido)acetic acid
Synonyms: (4-methylbenzenesulfonamido)acetic acid
Molecular Weight (g/mol): 229.25
Molecular Formula: C9H11NO4S
InChi Key: VDKFCCZUCXYILI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)