missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Tosylglycine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N-Tosylglycine |
| CAS | 1080-44-0 |
| Molecular Formula | C15 H14 Cl N O7 |
| Quantity | 5 g |
| Synonym | 2-(4-Methylphenylsulfonamido)acetic Acid, N-(p-Toluenesulfonyl)glycine, N-(p-Tolylsulfonyl)glycine, N-p-(Toluenesulfonyl)aminoacetic Acid, N-p-Tosylglycine, NSC 25821, Tosylglycine, [(Toluene-4-sulfonyl)amino]acetic Acid, [[(4-Methylphenyl)sulfonyl]amino]acetic Acid, N-[(4-Methylphenyl)sulfonyl]glycine, (p-Toluenesulfonyl)glycine |
| InChI Formula | InChI=1S/C15H14ClNO7/c16-7-3-4-8(9-6(7)2-1-5-17-9)23-15-12(20)10(18)11(19)13(24-15)14(21)22/h1-5,10-13,15,18-20H,(H,21,22)/t10-,11-,12+,13-,15+/m0/s1 |
| SMILES | O[C@H]1[C@H](Oc2ccc(Cl)c3cccnc23)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6S)-6-(5-chloroquinolin-8-yl)oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Molecular Weight (g/mol) | 355.727 |
| Formula Weight | 355.046 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction