accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNHClH2OHCl

CAS RN 1082699-15-7

IUPAC Name: 1-(2-chlorophenyl)piperazine hydrate hydrochloride
Synonyms: 1-(2-chlorophenyl)piperazine hydrate hydrochloride
Molecular Weight (g/mol): 251.15
Molecular Formula: C10H16Cl2N2O
InChi Key: RXPHNGRBTQWKEQ-UHFFFAOYSA-N
SMILES: O.Cl.ClC1=CC=CC=C1N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results