accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 108998-83-0

HOOH

CAS RN 108998-83-0

IUPAC Name: 1,1,2-triphenylethane-1,2-diol
Synonyms: 1,1,2-triphenylethane-1,2-diol
Molecular Weight (g/mol): 290.36
Molecular Formula: C20H18O2
InChi Key: GWVWUZJOQHWMFB-UHFFFAOYNA-N
SMILES: OC(C1=CC=CC=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results