accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NO

CAS RN 109-02-4

IUPAC Name: 4-methylmorpholine
Synonyms: N-methylmorpholine
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: SJRJJKPEHAURKC-UHFFFAOYSA-N
SMILES: CN1CCOCC1
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (4)
  • (2)
Boiling Point 
  • (1)
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (5)
  • (3)
  • (8)
  • (1)