accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 109-11-5

OOHN

CAS RN 109-11-5

IUPAC Name: morpholin-3-one
Synonyms: morpholin-3-one
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: VSEAAEQOQBMPQF-UHFFFAOYSA-N
SMILES: O=C1COCCN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results