accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 109-59-1

H3CCH3OOH

CAS RN 109-59-1

IUPAC Name: 2-(propan-2-yloxy)ethan-1-ol
Synonyms: 2-isopropoxyethanol
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: HCGFUIQPSOCUHI-UHFFFAOYSA-N
SMILES: CC(C)OCCO
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)