accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3HOCH3CH3HO

CAS RN 110-03-2

IUPAC Name: 2,5-dimethylhexane-2,5-diol
Synonyms: 2,5-hexanediol, 2,5-dimethyl-
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: ZWNMRZQYWRLGMM-UHFFFAOYSA-N
SMILES: CC(C)(O)CCC(C)(C)O
Boiling Point 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)