accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3CH3

CAS RN 110-27-0

IUPAC Name: propan-2-yl tetradecanoate
Synonyms: isopropyl myristate caswell no. 511E bisomel crodamol I.P.M. epa pesticide chemical code 000207 emerest 2314 wickenol 101 kessco ipm kessco isopropyl myristate ja-fa ipm
Molecular Weight (g/mol): 270.46
Molecular Formula: C17H34O2
InChi Key: AXISYYRBXTVTFY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)OC(C)C
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)