accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 110-63-4

IUPAC Name: butane-1,4-diol
Synonyms: 1,4-butanediol 1,4-butylene glycol
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: WERYXYBDKMZEQL-UHFFFAOYSA-N
SMILES: OCCCCO
Boiling Point 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)